Learn More
Thomas Scientific X-Pierce Film, Precut Vinyl, Sterile
SDP
Catalog No. NC9701030
Encompass
Precut Pierceable Sealing Films for Automation Protect samples and limit evaporation, short-term Temperatures -40°C to 90°C Special hairline X-cuts and thin adhesive prevent fouling or occlusion of probes and tips Sealing film reseals for continued sample protection after sampling X-Pierce sealing films are 100 µm thick vinyl with a 20 µm thick adhesive layer, designed for temporary protection from contamination and evaporation for samples in 96-well plates. A precut “X” over each well creates four flaps that easily bend downward when pushed by a robotic probe or pipet tip, allowing sample access without coring or adhesive fouling. A thin adhesive layer and special hairline cuts separating the X-Pierce flaps combine to prevent probes from contacting adhesive and becoming occluded. The flaps return to their original position after sampling for continued protection. Dimension are 145.5 x 79.4 mm. For long-term sample protection after sampling, a continuous
Catalog No. NC9701030 Supplier Thomas Scientific Supplier No. 1230Y22
Precut Pierceable Sealing Films for Automation Protect samples and limit evaporation, short-term Temperatures -40°C to 90°C Special hairline X-cuts and thin adhesive prevent fouling or occlusion of probes and tips Sealing film reseals for continued sample protection after sampling X-Pierce sealing films are 100 µm thick vinyl with a 20 µm thick adhesive layer, designed for temporary protection from contamination and evaporation for samples in 96-well plates. A precut “X” over each well creates four flaps that easily bend downward when pushed by a robotic probe or pipet tip, allowing sample access without coring or adhesive fouling. A thin adhesive layer and special hairline cuts separating the X-Pierce flaps combine to prevent probes from contacting adhesive and becoming occluded. The flaps return to their original position after sampling for continued protection. Dimension are 145.5 x 79.4 mm. For long-term sample protection after sampling, a continuous