Learn More
Draeger X-plore™ 1350 N95 Respirator
Catalog No. 17010972 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Size:
Medium/Large
Small/Medium
X-Small
With Exhalation Valve:
No
Yes
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size With Exhalation Valve
17-010-972 Medium/Large Yes
17-010-968 X-Small No
17-010-969 Small/Medium No
17-010-971 Small/Medium Yes
17-010-970 Medium/Large No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
WARNING. Air purifying respirators will help protect you in contaminated areas only if you strictly follow all the instructions and limitations on use set forth in the IFU. Failure to do so could lead to sickness or death!
Catalog No. 17-010-972 Supplier Draeger Supplier No. 3951356
  • $565.20
    $477.00 / Case of 180
    Save $88.20
    16% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
WARNING. Air purifying respirators will help protect you in contaminated areas only if you strictly follow all the instructions and limitations on use set forth in the IFU. Failure to do so could lead to sickness or death!
An ergonomic mask body combined with a secure, integrated nose seal and nose clip
  • CoolSAFE™ filter material produced from high-performance filter media combines low breathing resistance and high filter performance
  • Flexible nose clip ensures a great fit in the most critical area
  • VarioFLEX™ head harness with EasyStop adjustment for a pressure-free fit
  • CoolMAX exhalation valve (optional) effectively releases humid and warm exhaled air to the outside
  • Three sizes (XS, S/M and M/L) for an optimal fit for virtually every face
Specifications
Aerosol Type N - Not Resistant to Oil
Assigned Protection Factor (APF) 10
Certifications (NIOSH) Yes
Color White
Design Type Cup/Molded
Disposable Limited-use
Filter Type FFP2
Material Polypropylene
Product Type General Purpose Particulate Respirator
Size Medium/Large
With Exhalation Valve Yes
Latex Free Yes
Nose Piece Material Synthetic Foam