Learn More
Draeger X-plore™ 8000 Flexible Hose for PAPR
Catalog No. 17010993 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Size:
One Size
Catalog No. Size
17-010-993 One Size
1 options
WARNING. Air purifying respirators will help protect you in contaminated areas only if you strictly follow all the instructions and limitations on use set forth in the IFU. Failure to do so could lead to sickness or death!
Catalog No. 17-010-993 Supplier Draeger Supplier No. R59600
Add to Cart
Add to Cart
WARNING. Air purifying respirators will help protect you in contaminated areas only if you strictly follow all the instructions and limitations on use set forth in the IFU. Failure to do so could lead to sickness or death!
Ensures a continuous supply of clean air to the wearer's headpiece
  • Connects the Draeger headpiece to the X-plore™ 8000 series PAPR blower
  • Made of flexible material to provide freedom of motion
  • NIOSH-approved
Specifications
Color Black
Disposable Reusable
Material EPDM Rubber
Product Type Breathing Tube
Size One Size