Learn More
Draeger X-plore™ 8000 PAPR Helmets
Catalog No. 17010966 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Size:
One Size
Standard
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-010-966 One Size
17-010-959 Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
WARNING. Air purifying respirators will help protect you in contaminated areas only if you strictly follow all the instructions and limitations on use set forth in the IFU. Failure to do so could lead to sickness or death!
Catalog No. 17-010-966 Supplier Draeger Supplier No. 3729120
Add to Cart
Add to Cart
WARNING. Air purifying respirators will help protect you in contaminated areas only if you strictly follow all the instructions and limitations on use set forth in the IFU. Failure to do so could lead to sickness or death!
Helmets made for use in the harsh environment of day-to-day industrial operations
  • Optimal ANSI eye protection with a wide field of vision
  • Offer head and face protection against mechanical impacts, and some models also protect against chemical exposure
  • Durable and protective ANSI-rated helmet
  • Easily adjustable to any head size
  • Adjustable air outlet for a comfortable fit
  • Flip-up visor for enhanced worker communications
  • APR 25 and 1000
Specifications
ANSI Standard Yes
Assigned Protection Factor (APF) 1000
Certifications (NIOSH) Yes
Color Black
Disposable Reusable
Filter Type None
Material Nylon
Product Type Respirator Helmet
Size One Size
With Exhalation Valve No