Learn More
Draeger X-plore™ 8000 Premium PAPR Hood
Catalog No. 17010997 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Size:
Large/X-Large
Catalog No. Size
17-010-997 Large/X-Large
1 options
WARNING. Air purifying respirators will help protect you in contaminated areas only if you strictly follow all the instructions and limitations on use set forth in the IFU. Failure to do so could lead to sickness or death!
Catalog No. 17-010-997 Supplier Draeger Supplier No. R59870
Add to Cart
Add to Cart
WARNING. Air purifying respirators will help protect you in contaminated areas only if you strictly follow all the instructions and limitations on use set forth in the IFU. Failure to do so could lead to sickness or death!
The hood series offers the highest level of safety and has an optimised design that is comfortable even during extended use
  • Defined lens curvature with a robust PC lens
  • Hoods made from soft, low-noise material
  • Available in different sizes and lenghts
  • Choice of limited-use Standard model or durable Premium model
  • APF 1000
  • Recommended for use in all areas where no additional mechanical head or eye protection is needed
Specifications
Assigned Protection Factor (APF) 1000
Certifications (NIOSH) Yes
Color Gray
Disposable Limited-use
Filter Type None
Material Polypropylene
Product Type Double Bib Hood
Size Large/X-Large
With Exhalation Valve No
Latex Free Yes
Material (Lens) Polycarbonate
Model 8000 P
Sterility Non-sterile
Style Long
Dimensions (L x W x H) 10 x 9 x 19 cm