Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Draeger X-plore™ 8000 Series PAPR Application Set

Catalog No. 17010958 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Size:
Standard
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-010-958 Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING. Air purifying respirators will help protect you in contaminated areas only if you strictly follow all the instructions and limitations on use set forth in the IFU. Failure to do so could lead to sickness or death!
Catalog No. 17-010-958 Supplier Draeger Supplier No. 3703441
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING. Air purifying respirators will help protect you in contaminated areas only if you strictly follow all the instructions and limitations on use set forth in the IFU. Failure to do so could lead to sickness or death!

Fully integrated application sets, customizable respiratory protection kits built around the Draeger X-plore™ 8000 series (such as the 8500 or 8700 blower units)

  • Application sets for the Draeger X-plore™ 8500 or X-plore™ 8700 blowers
  • Bundled part number for core components of the PAPR system
  • Refer to the individual part numbers for guidance on additional components needed
  • NIOSH-approved

Specifications

Assigned Protection Factor (APF) 1000
Battery Type Lithium-ion
Belt Material Nylon
Certifications (NIOSH) Yes
Color Black
Product Type PAPR Assembly with Hood
Size Standard
Facepiece Material Polypropylene, Polyethylene
Facepiece Type Hood
Intrinsic Safety (IS) Rating No
IP Rating IP65
Material (Faceshield) Polycarbonate
Style Short
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.