Learn More
Dräger™ X-plore™ Multi-Gas+ Cartridges
Catalog No. 17625300 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
For Use With (Equipment):
X-plore 3300/3500/5500 twin-filter respirator
Catalog No. For Use With (Equipment)
17-625-300 X-plore 3300/3500/5500 twin-filter respirator
1 options
Catalog No. 17-625-300 Supplier Dräger™ Supplier No. 6738361
  • $1,959.00
    $1,301.65 / Case of 56
    Save $657.35
    34% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
For protection against: Organic Vapors, Chloride, Hydrogen Chloride, Sulfur Dioxide, Chlorine Dioxide, Hydrogen Fluoride, Formaldehyde, Ammonia, Methylamine, Hydrogen Sulfide
Fits Dräger X-plore 3300/3500 half masks and Dräger X-plore 5500full face masks
  • Strong blue ABS plastic housing
  • Safe, quick and easy bayonet connection with clear guidance bringing the filter/cartridge securely into place
  • Both filters are inserted in a top-down position
  • For use under visors high level of user safety
  • Option to upgrade a gas cartridge to a combination cartridge by simply adding a pad retrainer and a N 95 or R 95 pad
  • Shelflife (from date of production): gas and combination cartridges 6 years, pads 3 years
Order Info
Packaged in pairs for optimum protection of the unused filter. Instructions included with each pair.
Certifications
NIOSH 42 CFR 84
Specifications
Cartridge Type Multi-gas Cartridge
Certifications/Compliance NIOSH
Product Type Cartridge
For Use With (Application) For use under visors high level of user safety
For Use With (Equipment) X-plore 3300/3500/5500 twin-filter respirator
Quantity 1 Ea.