Learn More
Alkali Scientific X-Punch™ Pierceable Sealing Film, Sterile, 50/pk
SDP
Catalog No. 502388914
Encompass_Preferred
X-Punch™ pierceable sealing film is designed for short-term processing and storage of samples that require multiple access to wells by pipet tips or robotic probes. A pre-cut “X” over each well creates bendable flaps that provide sample access without coring or adhesive fouling of tips or probes. X-Punch has printed circles aid in the alignment of the film to the plate. It also has printed alphanumerics identify each well of a 96-well plate. It is piercable and reseals after piercing for continued sample protection.
Catalog No. 50-238-8914 Supplier Alkali Scientific Supplier No. SF35MLS
X-Punch™ pierceable sealing film is designed for short-term processing and storage of samples that require multiple access to wells by pipet tips or robotic probes. A pre-cut “X” over each well creates bendable flaps that provide sample access without coring or adhesive fouling of tips or probes. X-Punch has printed circles aid in the alignment of the film to the plate. It also has printed alphanumerics identify each well of a 96-well plate. It is piercable and reseals after piercing for continued sample protection.