Learn More
Med Vet International Slimlite X-Ray Illuminator Accessories, 4-Amp Battery Pack for X-Ray Illuminator, J0806Q
Catalog No. 5000495545
Encompass_Preferred
High quality radiographs should be matched with a high quality viewing box for the maximum diagnostic image. The JorVet Slim Lite viewing box uses new "Edgelite" technology with proprietary light diffusion material giving an almost sunlight illumination with 5,000° Kelvin color temperature. In addition, this "Edgelite" format allows a much thinner or streamlined attractive profile than other viewing boxes.
Catalog No. 50-004-95545 Supplier Med Vet International Supplier No. JOR806Q
High quality radiographs should be matched with a high quality viewing box for the maximum diagnostic image. The JorVet Slim Lite viewing box uses new "Edgelite" technology with proprietary light diffusion material giving an almost sunlight illumination with 5,000° Kelvin color temperature. In addition, this "Edgelite" format allows a much thinner or streamlined attractive profile than other viewing boxes.