Learn More
Cole-Parmer Cole-Parmer Blue-Sensitive Autoradiography Film, 8 x 10", 100/pk
Catalog No. NC9977921 Shop All Cole-Parmer Products
Encompass
Ideal for western blotting, DNA sequencing gels, chemiluminescent probes, gel shift analysis, and commonly used radioisotopes (32P, 125I). Use for manual or automatic development.
Catalog No. NC9977921 Supplier Cole-Parmer Supplier No. 2170003
Ideal for western blotting, DNA sequencing gels, chemiluminescent probes, gel shift analysis, and commonly used radioisotopes (32P, 125I). Use for manual or automatic development.