Learn More
Research Products International Corp X-Ray Cassette Security Bag for 5 x 7 Inch Cassette
SDP
Catalog No. 50212932
Encompass
Heavy duty black fabric bags provide an extra measure of protection against light leakage. Especially useful when using cassettes that may be worn or slightly damaged from use. Ideal for preventing cassettes from sticking together when stacked in a freezer. Bags have a full width sealing strip.
Catalog No. 50-212-932 Supplier Research Products International Corp Supplier No. F133530057
Heavy duty black fabric bags provide an extra measure of protection against light leakage. Especially useful when using cassettes that may be worn or slightly damaged from use. Ideal for preventing cassettes from sticking together when stacked in a freezer. Bags have a full width sealing strip.