Learn More
Med Vet International X-Ray Developing Hanger, 8in X 10in, J0678
Catalog No. 5000495934
Encompass_Preferred
Stainless steel metal hanger holds the x-ray film while in the developing or processing tanks. Clip on to the four corners of the film.
Catalog No. 50-004-95934 Supplier Med Vet International Supplier No. JOR678
Stainless steel metal hanger holds the x-ray film while in the developing or processing tanks. Clip on to the four corners of the film.