Learn More
Med Vet International X Ray Markers, (Left and Right), J1185, Set of 2
Catalog No. 5000495703
Encompass_Preferred
This set of two x-ray markers (L and R) does more than give the side orientation of an x-ray. It has a cm measurement scale. This is useful for sending digital x-rays to other parties that may have a different software or type of digital radiography. Markers are 50mm long and marked in 5mm intervals. INITIALS CANNOT BE CHANGED
Catalog No. 50-004-95703 Supplier Med Vet International Supplier No. JOR1185
This set of two x-ray markers (L and R) does more than give the side orientation of an x-ray. It has a cm measurement scale. This is useful for sending digital x-rays to other parties that may have a different software or type of digital radiography. Markers are 50mm long and marked in 5mm intervals. INITIALS CANNOT BE CHANGED