Learn More
Med Vet International X-Ray Measuring Caliper, J0684, Each
Supplier: Med Vet International JOR684
Getting the correct thickness of your patient is critical to proper KVP setting and quality diagnostic radiographs. Measurements in centimeters only. Stainless steel.