Learn More
Med Vet International X-Ray Protective Gloves, J0677X, 1 Pair
Catalog No. 5000495392
Encompass_Preferred
Give consistent radiation protection with better finger and hand flexibility. Seamless lead vinyl gloves are a 14"" one-piece dip molded glove with supple vinyl cover and foam liner. Burgundy color. Standard protection 0.5mm Pb equivalency
Catalog No. 50-004-95392 Supplier Med Vet International Supplier No. JOR677X
Add to Cart
Add to Cart
Give consistent radiation protection with better finger and hand flexibility. Seamless lead vinyl gloves are a 14"" one-piece dip molded glove with supple vinyl cover and foam liner. Burgundy color. Standard protection 0.5mm Pb equivalency