Learn More
Med Vet International Wrap Around Protective X-Ray Eyewear, J0676E, Each
Catalog No. 5000495985
Encompass_Preferred
Features molded wrap styling in durable tortoise nylon frame with leaded side shields for added protection. Standard protection: front & side lenses 0.75mm Pb equivalent optical lead glass
Catalog No. 50-004-95985 Supplier Med Vet International Supplier No. JOR676E
Features molded wrap styling in durable tortoise nylon frame with leaded side shields for added protection. Standard protection: front & side lenses 0.75mm Pb equivalent optical lead glass