Learn More
Electron Microscopy Sciences X-Ray Window Silicon Nitride Film, 1000 µm, 50 nm
SDP
Catalog No. 501929443
Encompass_Preferred
Ideal substrates for x-ray microscopy and x-ray spectroscopy techniques. State-of-the-art manufacturing and expert engineering allow for competitive prices of these windows. Made in the USA, these X-Ray windows are entirely plasma cleanable to remove organic contamination. Flat, uniformly deposited films provide consistent backgrounds with low field-to-field variability and high x-ray transmission. For high temperature and differential pressure environments.
Dimensions: 1000 µm
Catalog No. 50-192-9443 Supplier Electron Microscopy Sciences Supplier No. 7604213
Ideal substrates for x-ray microscopy and x-ray spectroscopy techniques. State-of-the-art manufacturing and expert engineering allow for competitive prices of these windows. Made in the USA, these X-Ray windows are entirely plasma cleanable to remove organic contamination. Flat, uniformly deposited films provide consistent backgrounds with low field-to-field variability and high x-ray transmission. For high temperature and differential pressure environments.
Dimensions: 1000 µm