Learn More
Electron Microscopy Sciences X-Ray Window Silicon Nitride Film, 500 µm, 100 nm
SDP
Catalog No. 501929444
Encompass_Preferred
Ideal substrates for x-ray microscopy and x-ray spectroscopy techniques. State-of-the-art manufacturing and expert engineering allow for competitive prices of these windows. Made in the USA, these X-Ray windows are entirely plasma cleanable to remove organic contamination. Flat, uniformly deposited films provide consistent backgrounds with low field-to-field variability and high x-ray transmission. For high temperature and differential pressure environments.
Dimensions: 500 µm
Catalog No. 50-192-9444 Supplier Electron Microscopy Sciences Supplier No. 7604214
Ideal substrates for x-ray microscopy and x-ray spectroscopy techniques. State-of-the-art manufacturing and expert engineering allow for competitive prices of these windows. Made in the USA, these X-Ray windows are entirely plasma cleanable to remove organic contamination. Flat, uniformly deposited films provide consistent backgrounds with low field-to-field variability and high x-ray transmission. For high temperature and differential pressure environments.
Dimensions: 500 µm