Learn More
Chemglass Life Sciences Chemglass X-Shaped PTFE Magnetic Stir Bar, 10 x 5 mm, Pack of 100 - For Magnetic Stirrers
SDP
Catalog No. NC2254927
Encompass_Preferred
Chemglass X-shaped PTFE magnetic stir bars (10 x 5 mm) are designed to create a deep vortex and provide stable stirring in laboratory vessels. Supplied bulk-packed in quantities of 100 (P100), these PTFE-coated stir bars are manufactured with FDA-grade, USP Class VI materials and are suitable for routine mixing on magnetic stirrers.
  • 10 x 5 mm (diameter x height)
  • PTFE construction, FDA-grade and USP Class VI
  • X-shaped design creates a deep vortex and improves stability during stirring
  • Bulk packed - 100 per pack (P100)
  • Designed for use with magnetic stirrers
This item is not returnable. View return policy
Catalog No. NC2254927 Supplier Chemglass Life Sciences Supplier No. CG2013101
Chemglass X-shaped PTFE magnetic stir bars (10 x 5 mm) are designed to create a deep vortex and provide stable stirring in laboratory vessels. Supplied bulk-packed in quantities of 100 (P100), these PTFE-coated stir bars are manufactured with FDA-grade, USP Class VI materials and are suitable for routine mixing on magnetic stirrers.
  • 10 x 5 mm (diameter x height)
  • PTFE construction, FDA-grade and USP Class VI
  • X-shaped design creates a deep vortex and improves stability during stirring
  • Bulk packed - 100 per pack (P100)
  • Designed for use with magnetic stirrers