Learn More
Chemglass Life Sciences Chemglass X-Shaped PTFE Magnetic Stir Bar, 20 x 8 mm, Pack of 100
SDP
Catalog No. NC2254932
Encompass_Preferred
Chemglass X-shaped PTFE magnetic stir bars are bulk-packed stir bars designed for use with laboratory magnetic stirrers. Each bar measures 20 x 8 mm; the pack contains 100 pieces for bulk laboratory use.
  • Ptfe construction provides chemical resistance and broad solvent compatibility.
  • X-shaped profile creates a deep vortex and provides stability during stirring.
  • 20 x 8 mm size is suitable for small to medium vessels.
  • Pack of 100 supplies laboratories with a bulk quantity for routine use.
  • White color allows easy visibility in clear solutions.
This item is not returnable. View return policy
Catalog No. NC2254932 Supplier Chemglass Life Sciences Supplier No. CG2013102
Chemglass X-shaped PTFE magnetic stir bars are bulk-packed stir bars designed for use with laboratory magnetic stirrers. Each bar measures 20 x 8 mm; the pack contains 100 pieces for bulk laboratory use.
  • Ptfe construction provides chemical resistance and broad solvent compatibility.
  • X-shaped profile creates a deep vortex and provides stability during stirring.
  • 20 x 8 mm size is suitable for small to medium vessels.
  • Pack of 100 supplies laboratories with a bulk quantity for routine use.
  • White color allows easy visibility in clear solutions.