Learn More
Lomir Biomedical Inc X-SMALL RAT SNUGGLE
SDP
Catalog No. NC4010667
Encompass_Preferred
The “Snuggle Family” uses the principle of a cocoon rather than a rigid structure for immobilization. The multiple flaps perform two functions: they are adjustable around the subject for effective yet comfortable restraint and can be closed or left open for access depending on the procedure. Both restraint and procedure can be carried out by one handler, thus reducing stress experienced by the subject.
Catalog No. NC4010667 Supplier Lomir Biomedical Inc Supplier No. RTS 00
The “Snuggle Family” uses the principle of a cocoon rather than a rigid structure for immobilization. The multiple flaps perform two functions: they are adjustable around the subject for effective yet comfortable restraint and can be closed or left open for access depending on the procedure. Both restraint and procedure can be carried out by one handler, thus reducing stress experienced by the subject.