Description
- Sport-grip™ temples
- Ratchet adjustment for three lens angles
- 7.5 base lens is mounted in a lightweight nylon frame
- Provides maximum coverage and excellent peripheral vision
- Lens with DX™ anti-fog hard coat protects from fog, scratching, and most chemicals
- 3M Catalog# 15176-00000-20 , 3M Catalog# 15182-00000-20
Compliance
Meets the requirements of ANSI™ Z87.1-2010 and CSA Z94.3-2007.
Specifications
Specifications
| Certifications/Compliance | Meets the requirements of ANSI Z87.1-2010 and CSA Z94.3-2007. |
| Lens Tint | Clear |
| Frame Color | Silverized |
| Model | 15182-00000-20 |
| Temple Style | Sport Grip |
| Frame Material | Nylon |