Learn More
Sigma Aldrich Fine Chemicals Biosciences X-tremeGENE HP DNA Transfection Reagent, 0.4 mL
Catalog No. NC1116383
Encompass
X-tremeGENE HP DNA Transfection Reagent is a multi-component reagent that forms a complex with DNA , then transports the complex into animal or insect cells.
Catalog No. NC1116383 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 6366244001
X-tremeGENE HP DNA Transfection Reagent is a multi-component reagent that forms a complex with DNA , then transports the complex into animal or insect cells.