Learn More
Sigma Aldrich Fine Chemicals Biosciences X-tremeGENETM 360 Transfec
Catalog No. 502905347
Encompass
X-tremeGENE(TM) 360 Transfection Reagent is an innovative reagent that forms a complex with DNA or RNA and efficiently delivers the nucleic acids into animal or insect cells. With improved utility X-tremeGENE(TM) 360 delivers high efficiency transfection in more cell types with low cytotoxicity.
Catalog No. 50-290-5347 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 8724105001
X-tremeGENE(TM) 360 Transfection Reagent is an innovative reagent that forms a complex with DNA or RNA and efficiently delivers the nucleic acids into animal or insect cells. With improved utility X-tremeGENE(TM) 360 delivers high efficiency transfection in more cell types with low cytotoxicity.