Learn More
Sigma Aldrich Fine Chemicals Biosciences X tremeGENETM HP DNA Trans
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 6366236001
Encompass
X-tremeGENE HP DNA Transfection Reagent is a multi-component reagent that forms a complex with DNA then transports the complex into animal or insect cells.
Catalog No. 50-290-5184