Learn More
Lonza Walkersville X-VIVO 15 Serum-free Hematopoietic Cell Medium
Catalog No. NC2540740
Encompass
X-VIVOTM 15 is a high performing media that provides nutritionally adequate and balanced environments for a variety of hematopoietic cells including:T cellsNatural killer cells (NK)Peripheral blood lymphocytes (PBL)Tumor infiltrating lymphocytes (TIL)Dendritic cells (DC)Human monocytesMacrophage cellsX-VIVOTM 15 Media are mostly known for its ability to support the proliferation of purified CD3+ cells isolated from peripheral blood and human tumors making it well suited for CAR-T applications. THIS PRODUCT IS NOT FOR USE IN GMP MANUFACTURING, NOR HUMAN OR ANIMAL IN VIVO USE, INCLUDING USE AS A DILUENT OR AS AN EXCIPIENT, OR FOR DIAGNOSTIC USE. This product is for research use only.Applications:Proliferation of peripheral blood lymphocytes Proliferation of tumor infiltrating lymphocytes Cryopreservation of human tissue Cultivation of human monocytes and macrophages Cultivation of stem cells Cultivation of dendritic cells
Catalog No. NC2540740 Supplier Lonza Walkersville Supplier No. 02060Q
X-VIVOTM 15 is a high performing media that provides nutritionally adequate and balanced environments for a variety of hematopoietic cells including:T cellsNatural killer cells (NK)Peripheral blood lymphocytes (PBL)Tumor infiltrating lymphocytes (TIL)Dendritic cells (DC)Human monocytesMacrophage cellsX-VIVOTM 15 Media are mostly known for its ability to support the proliferation of purified CD3+ cells isolated from peripheral blood and human tumors making it well suited for CAR-T applications. THIS PRODUCT IS NOT FOR USE IN GMP MANUFACTURING, NOR HUMAN OR ANIMAL IN VIVO USE, INCLUDING USE AS A DILUENT OR AS AN EXCIPIENT, OR FOR DIAGNOSTIC USE. This product is for research use only.Applications:Proliferation of peripheral blood lymphocytes Proliferation of tumor infiltrating lymphocytes Cryopreservation of human tissue Cultivation of human monocytes and macrophages Cultivation of stem cells Cultivation of dendritic cells