Learn More
Lonza Walkersville X-VIVO 20 HSC Cell Medium
Catalog No. 5000496418
Encompass
X-VIVO 20 Medium is optimized to support the generation of lymphokine activated killer (LAK) cells from monocytedepleted peripheral blood lymphocytes (PBL) at high density Initial cell densities between 2 0 - 3 0 10 7 cells/ ml can be used to successfully generate LAK cells X-VIVO 20 Medium may also be used as a growth medium for PBL and tumor infiltrating lymphocytes (TIL) All BioWhittaker Cell Culture Media products are for Research Use Only (RUO) and are not approved for human or veterinary use or for use in clinical or in vitro diagnostic procedures If you require GMP grade media contact Lonza for more details
This item is not returnable. View return policy
Catalog No. 50-004-96418 Supplier Lonza Walkersville Supplier No. 04448Q
X-VIVO 20 Medium is optimized to support the generation of lymphokine activated killer (LAK) cells from monocytedepleted peripheral blood lymphocytes (PBL) at high density Initial cell densities between 2 0 - 3 0 10 7 cells/ ml can be used to successfully generate LAK cells X-VIVO 20 Medium may also be used as a growth medium for PBL and tumor infiltrating lymphocytes (TIL) All BioWhittaker Cell Culture Media products are for Research Use Only (RUO) and are not approved for human or veterinary use or for use in clinical or in vitro diagnostic procedures If you require GMP grade media contact Lonza for more details