Learn More
Montana Molecular Green Gi cADDis cAMP sensor Big Sky BacMam 5mL
SDP
Catalog No. 502815251
Encompass_Preferred
Detect cAMP Gi signaling kinetics in living cells with the Green Gi cADDis fluorescent cAMP assay. Packaged in Big Sky BacMam, for enhanced expression in HEKs, CHOs, and many other mammalian cell types and iPSCs. This product is an update to product X0200G, with improved expression efficiency from Big Sky BacMam.
Catalog No. 50-281-5251 Supplier Montana Molecular Supplier No. X0250G005
Detect cAMP Gi signaling kinetics in living cells with the Green Gi cADDis fluorescent cAMP assay. Packaged in Big Sky BacMam, for enhanced expression in HEKs, CHOs, and many other mammalian cell types and iPSCs. This product is an update to product X0200G, with improved expression efficiency from Big Sky BacMam.