Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex X60, 34770 Wypall Dispenser
SDP

Catalog No. 01259221
Change view
Click to view available options
Mounting:
Magnetic Mount
VHB Tape
Wall Mount
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
01-259-221 Wall Mount
01-259-195 Magnetic Mount
01-259-158 VHB Tape
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
N/A
Catalog No. 01-259-221 Supplier Poltex Supplier No. WYPL34770W
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N/A

Holds and dispenses Wypall X60 34770 wipes. Choose from Wall Mount, HB Tape Mount, or Magnet Mount with durable PETG construction.

  • Holds and dispenses Wypall X60, 34770 wipes.
  • Choose from Wall Mount (screws/anchors included), HB Tape Mount (high-bond tape included), or Magnet Mount (powerful magnets included).
  • Durable and cleanable PETG construction.
  • Consumables not included.

Specifications

Product Type Wipe Dispenser
Color Clear
Dimensions (L x W x H) 6.2 x 12.5 x 7.5 in.
Holds Labwipes
Material PETG
Mounting Wall Mount
No. of Compartments 1
Product Style Open Top and Front
Edges Rounded
For Use With (Application) Holds and dispenses Wypall
Includes Mounting Hardware
Quantity 1 Ea.
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.