Learn More
Fluidigm Corp X8 ANTIBODY LABELING KIT 151EU
SDP
Supplier:  Fluidigm Corp 201151A
Encompass
This Maxpar kit lets you conjugate IgG antibodies with the 151eu metal isotope, with each kit containing enough material to conjugate 440 antibodies in 100 g amounts.
Catalog No. NC0269333