Learn More
Electron Microscopy Sciences Omniprobe XA Probe Point Holder
SDP
Catalog No. 5033760
Encompass
AutoProbe 250 probe tip holder uses the tungsten probe tip75960-05. Compatible with the Short-Cut for direct conversion to TEM Grid. Comes with a storage vial and handle.
Catalog No. 50-337-60 Supplier Electron Microscopy Sciences Supplier No. 7596110
AutoProbe 250 probe tip holder uses the tungsten probe tip75960-05. Compatible with the Short-Cut for direct conversion to TEM Grid. Comes with a storage vial and handle.