Learn More
Apexbio Technology LLC XAC 96865-92-8 5mg
SDP
Catalog No. 502240478
Encompass_Preferred
XAC is a small-molecule antagonist targeting adenosine receptors It is designed to competitively inhibit the binding of adenosine to its receptors thereby modulating receptor-mediated signaling pathways XAC exerts its biological activity primarily through interference with adenosine-induced signaling Experimental studies indicate that XAC inhibits adenosine-induced cellular processes such as neurotransmission modulation and cardiovascular responses Based on these pharmacological properties XAC holds research potential in neuroscience and cardiovascular physiology to investigate adenosine-mediated signaling pathways in specific cellular events and physiological conditions
This item is not returnable. View return policy
Catalog No. 50-224-0478 Supplier Apexbio Technology LLC Supplier No. B73145
XAC is a small-molecule antagonist targeting adenosine receptors It is designed to competitively inhibit the binding of adenosine to its receptors thereby modulating receptor-mediated signaling pathways XAC exerts its biological activity primarily through interference with adenosine-induced signaling Experimental studies indicate that XAC inhibits adenosine-induced cellular processes such as neurotransmission modulation and cardiovascular responses Based on these pharmacological properties XAC holds research potential in neuroscience and cardiovascular physiology to investigate adenosine-mediated signaling pathways in specific cellular events and physiological conditions