Learn More
MSC X-Acto Knife Set, No. 1, 5 No. 11, 6 pieces
Catalog No. 5000685807
Encompass
X-Acto all-purpose knives have been a long-standing tool for artists, crafters, hobbyists, designers, and engineers due to their precision, durability, reliability, and detailed capabilities. These knives are designed to meet the needs of meticulous artists, featuring comfortable handles and strong, yet flexible, carbon steel blades.
  • Classic precision cutting tool
  • Aluminum handle with textured grip
  • Includes carbon steel blade and 5 replacement #11 blades
  • Versatile for trimming and cutting materials such as paper, cardstock, foam board, plastic, fabric, and thin metal
Catalog No. 50-006-85807 Supplier MSC Supplier No. 62809728
X-Acto all-purpose knives have been a long-standing tool for artists, crafters, hobbyists, designers, and engineers due to their precision, durability, reliability, and detailed capabilities. These knives are designed to meet the needs of meticulous artists, featuring comfortable handles and strong, yet flexible, carbon steel blades.
  • Classic precision cutting tool
  • Aluminum handle with textured grip
  • Includes carbon steel blade and 5 replacement #11 blades
  • Versatile for trimming and cutting materials such as paper, cardstock, foam board, plastic, fabric, and thin metal