Learn More
Aviva Systems Biology Corp XAF1 Antibody - middle region (ARP56970_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56970_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against XAF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human XAF1. Alternate names: BIRC4BP, HSXIAPAF1, XIAPAF1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-62048