Learn More
Aviva Systems Biology Corp XAGE2 Antibody - middle region (ARP59905_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59905_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against XAGE2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human XAGE2. Alternate names: CT12.2, GAGED3, XAGE-2, XAGE2BApplicationWB. Species reactivity: Human
Catalog No. 50-286-872