Learn More
Medchemexpress LLC Xaliproden hydrochloride | 90494-79-4 | 98.4% | 417.89 | 1 ML
SDP
Catalog No. 5000364049
Encompass_Preferred
Xaliproden hydrochloride (SR57746A) is a potent, selective, and orally active agonist of the 5-HT1A receptor, showing high affinity for 5-HT1A specific binding sites in the rat hippocampus (IC50=3 nM). It is also a selective antagonist of the dopamine D2 receptor, with moderate affinity (IC50=0.1-1 μM). Xaliproden hydrochloride exhibits anti-depression and anti-anxiety effects and may have therapeutic potential for the research of neurodegenerative diseases.
  • Potent, selective, and orally active 5-HT1A receptor agonist
  • High affinity for 5-HT1A specific binding sites (IC50=3 nM)
  • Selective antagonist of the dopamine D2 receptor
  • Moderate affinity for D2 receptor (IC50=0.1-1 μM)
  • Exhibits anti-depression and anti-anxiety effects
  • May have therapeutic potential for neurodegenerative disease research
Catalog No. 50-003-64049 Supplier Medchemexpress LLC Supplier No. HY1460410MM/1ML
Xaliproden hydrochloride (SR57746A) is a potent, selective, and orally active agonist of the 5-HT1A receptor, showing high affinity for 5-HT1A specific binding sites in the rat hippocampus (IC50=3 nM). It is also a selective antagonist of the dopamine D2 receptor, with moderate affinity (IC50=0.1-1 μM). Xaliproden hydrochloride exhibits anti-depression and anti-anxiety effects and may have therapeutic potential for the research of neurodegenerative diseases.
  • Potent, selective, and orally active 5-HT1A receptor agonist
  • High affinity for 5-HT1A specific binding sites (IC50=3 nM)
  • Selective antagonist of the dopamine D2 receptor
  • Moderate affinity for D2 receptor (IC50=0.1-1 μM)
  • Exhibits anti-depression and anti-anxiety effects
  • May have therapeutic potential for neurodegenerative disease research