Learn More
Medchemexpress LLC Xaliproden hydrochloride | 90494-79-4 | 98.4% | 417.89 | 100 MG
SDP
Catalog No. 5000363561
Encompass_Preferred
Xaliproden hydrochloride is a potent, selective, and orally active agonist of the 5-HT1A receptor. It also acts as a selective antagonist of the dopamine D2 receptor. This compound exhibits anti-depression and anti-anxiety effects and may have therapeutic potential for research into neurodegenerative diseases.
  • Potent, selective, and orally active agonist of 5-HT1A receptor
  • Selective antagonist of dopamine D2 receptor
  • Exhibits anti-depression and anti-anxiety effects
  • May have therapeutic potential for research into neurodegenerative diseases
Catalog No. 50-003-63561 Supplier Medchemexpress LLC Supplier No. HY14604100MG
Xaliproden hydrochloride is a potent, selective, and orally active agonist of the 5-HT1A receptor. It also acts as a selective antagonist of the dopamine D2 receptor. This compound exhibits anti-depression and anti-anxiety effects and may have therapeutic potential for research into neurodegenerative diseases.
  • Potent, selective, and orally active agonist of 5-HT1A receptor
  • Selective antagonist of dopamine D2 receptor
  • Exhibits anti-depression and anti-anxiety effects
  • May have therapeutic potential for research into neurodegenerative diseases