Learn More
Medchemexpress LLC Xaliproden hydrochloride | 90494-79-4 | 98.4% | 417.89 | 50 MG
SDP
Catalog No. 5000362600
Encompass_Preferred
Xaliproden hydrochloride (SR57746A) is a potent, selective, and orally active agonist of the 5-HT1A receptor. It exhibits high affinity for 5-HT1A specific binding sites in the rat hippocampus and acts as a selective antagonist of the dopamine D2 receptor. This compound demonstrates anti-depression and anti-anxiety effects.
  • Potent and selective 5-HT1A receptor agonist
  • Orally active
  • High affinity for 5-HT1A specific binding sites (IC50=3 nM)
  • Selective antagonist of dopamine D2 receptor (IC50=0.1-1 μM)
  • Potential therapeutic agent for neurodegenerative diseases research
Catalog No. 50-003-62600 Supplier Medchemexpress LLC Supplier No. HY1460450MG
Xaliproden hydrochloride (SR57746A) is a potent, selective, and orally active agonist of the 5-HT1A receptor. It exhibits high affinity for 5-HT1A specific binding sites in the rat hippocampus and acts as a selective antagonist of the dopamine D2 receptor. This compound demonstrates anti-depression and anti-anxiety effects.
  • Potent and selective 5-HT1A receptor agonist
  • Orally active
  • High affinity for 5-HT1A specific binding sites (IC50=3 nM)
  • Selective antagonist of dopamine D2 receptor (IC50=0.1-1 μM)
  • Potential therapeutic agent for neurodegenerative diseases research