Learn More
Medchemexpress LLC Xaliproden hydrochloride | 90494-79-4 | 98.6% | 417.89 g/mol | C24H23ClF3N | 5 MG
SDP
Catalog No. 5000221140
Encompass_Preferred
Xaliproden hydrochloride is the hydrochloride salt of a small-molecule 5-HT1A receptor agonist used in preclinical pharmacology and neuropharmacology research. It shows neurotrophic and neuroprotective effects in vitro and in vivo and is supplied for laboratory use in solid form and as DMSO solutions.
  • Acts as a potent, selective 5-HT1A receptor agonist.
  • Exhibits neurotrophic and neuroprotective activity in preclinical models.
  • Offered as solid and as 10 mM solution in DMSO for flexible use.
  • Provided as hydrochloride salt to improve handling and solubility.
  • High purity appropriate for research applications.
Catalog No. 50-002-21140 Supplier Medchemexpress LLC Supplier No. HY146045MG
Xaliproden hydrochloride is the hydrochloride salt of a small-molecule 5-HT1A receptor agonist used in preclinical pharmacology and neuropharmacology research. It shows neurotrophic and neuroprotective effects in vitro and in vivo and is supplied for laboratory use in solid form and as DMSO solutions.
  • Acts as a potent, selective 5-HT1A receptor agonist.
  • Exhibits neurotrophic and neuroprotective activity in preclinical models.
  • Offered as solid and as 10 mM solution in DMSO for flexible use.
  • Provided as hydrochloride salt to improve handling and solubility.
  • High purity appropriate for research applications.