Learn More
TARGETMOL CHEMICALS INC Xamoterol hemifumarate 10MG
SDP
Catalog No. 502877723
Encompass_Preferred
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Xamoterol hemifumarate is a potent and selective beta1-adrenoceptor agonist.Xamoterol hemifumarate is a potential compound for the study of the relationship between cardiac arrhythmias and beta1-adrenergic stimulation with IKr. Purity 99.79%
Catalog No. 50-287-7723 Supplier TARGETMOL CHEMICALS INC Supplier No. T2353610MG
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Xamoterol hemifumarate is a potent and selective beta1-adrenoceptor agonist.Xamoterol hemifumarate is a potential compound for the study of the relationship between cardiac arrhythmias and beta1-adrenergic stimulation with IKr. Purity 99.79%