Learn More
Medchemexpress LLC Xamoterol hemifumarate | 73210-73-8 | MFCD00661103 | 99.9% | 397.43 g/mol | C20H29N3O9 | 50 MG
SDP
Catalog No. 5000211506
Encompass_Preferred
Xamoterol hemifumarate is a selective, potent beta1-adrenergic receptor agonist used as a research reagent to study arrhythmogenesis and the relationship between β1-adrenergic stimulation and IKr.
  • Selective beta1-adrenergic receptor agonist.
  • Suitable for research into arrhythmogenesis and cardiac electrophysiology.
  • Available in small solid quantities, including 50 MG.
  • High purity (≈99.9%).
  • Suitable for sealed, low-temperature storage.
  • Includes CAS number 73210-73-8 for substance identification.
Catalog No. 50-002-11506 Supplier Medchemexpress LLC Supplier No. HY101327A50MG
Xamoterol hemifumarate is a selective, potent beta1-adrenergic receptor agonist used as a research reagent to study arrhythmogenesis and the relationship between β1-adrenergic stimulation and IKr.
  • Selective beta1-adrenergic receptor agonist.
  • Suitable for research into arrhythmogenesis and cardiac electrophysiology.
  • Available in small solid quantities, including 50 MG.
  • High purity (≈99.9%).
  • Suitable for sealed, low-temperature storage.
  • Includes CAS number 73210-73-8 for substance identification.