Learn More
Medchemexpress LLC Xamoterol hemifumarate | 73210-73-8 | MFCD00661104 | 99.9% | 397.43 g/mol | C20H29N3O9 | 100 MG
SDP
Catalog No. 5000211412
Encompass_Preferred
Xamoterol hemifumarate is a research-grade small molecule that acts as a selective and potent β1-adrenergic receptor agonist. It is supplied as the hemifumarate salt for laboratory research and is commonly used in pharmacology and cardiac studies requiring β1-selective stimulation.
  • Selective β1-adrenergic receptor agonist.
  • High stated purity (99.9%).
  • Supplied as a hemifumarate salt in a stable solid form.
  • Available in multiple milligram pack sizes, including 100 mg.
  • Suitable for in vitro pharmacology and cardiac research.
  • Provided with product documentation and certificate of analysis.
Catalog No. 50-002-11412 Supplier Medchemexpress LLC Supplier No. HY101327A100MG
Xamoterol hemifumarate is a research-grade small molecule that acts as a selective and potent β1-adrenergic receptor agonist. It is supplied as the hemifumarate salt for laboratory research and is commonly used in pharmacology and cardiac studies requiring β1-selective stimulation.
  • Selective β1-adrenergic receptor agonist.
  • High stated purity (99.9%).
  • Supplied as a hemifumarate salt in a stable solid form.
  • Available in multiple milligram pack sizes, including 100 mg.
  • Suitable for in vitro pharmacology and cardiac research.
  • Provided with product documentation and certificate of analysis.