Learn More
Cayman Chemical Xamoterol (hemifumarate), Purity: ≥98%, CAS: 73210-73-8, 10 mg
SDP
Catalog No. 502933433
Encompass_Preferred
A β1-adrenergic receptor partial agonist (EC50 = 80 nM for the generation of cAMP in neonatal rat cardiomyocyte cultures); increases spontaneous contraction of isolated rat right atria (EC50 = 4.67 nM); increases heart rate in beagle dogs (ED50 = 3.2 µg/kg) and a rat model of spontaneous heart failure (ED50 = 6 µg/kg)
Catalog No. 50-293-3433 Supplier Cayman Chemical Supplier No. 2426710 MG
A β1-adrenergic receptor partial agonist (EC50 = 80 nM for the generation of cAMP in neonatal rat cardiomyocyte cultures); increases spontaneous contraction of isolated rat right atria (EC50 = 4.67 nM); increases heart rate in beagle dogs (ED50 = 3.2 µg/kg) and a rat model of spontaneous heart failure (ED50 = 6 µg/kg)