Learn More
Medchemexpress LLC Xanomeline | 131986-45-3 | MFCD00867179 | 100.0% | 281.42 g·mol⁻¹ | C14H23N3OS | 100 MG
SDP
Catalog No. 5000244165
Encompass_Preferred
Xanomeline is a selective muscarinic M1/M4 receptor agonist used as a research compound to study cholinergic signaling, cognition, and neuropsychiatric disorders. Supplied as a solid (white to yellow), it has formula C14H23N3OS, molecular weight 281.42 g·mol⁻¹, and CAS 131986-45-3. It is intended for analytical and pharmacology applications in both in vitro and in vivo settings.
  • Selective M1/M4 receptor agonist for targeted pharmacology studies.
  • Suitable for in vitro and in vivo research applications.
  • High reported purity (99.95%).
  • Provided as a solid, white to yellow powder for ease of handling.
  • Molecular formula and weight included for analytical reference.
  • Available in multiple pack sizes for experimental flexibility.
Catalog No. 50-002-44165 Supplier Medchemexpress LLC Supplier No. HY105182100MG
Xanomeline is a selective muscarinic M1/M4 receptor agonist used as a research compound to study cholinergic signaling, cognition, and neuropsychiatric disorders. Supplied as a solid (white to yellow), it has formula C14H23N3OS, molecular weight 281.42 g·mol⁻¹, and CAS 131986-45-3. It is intended for analytical and pharmacology applications in both in vitro and in vivo settings.
  • Selective M1/M4 receptor agonist for targeted pharmacology studies.
  • Suitable for in vitro and in vivo research applications.
  • High reported purity (99.95%).
  • Provided as a solid, white to yellow powder for ease of handling.
  • Molecular formula and weight included for analytical reference.
  • Available in multiple pack sizes for experimental flexibility.