Learn More
Medchemexpress LLC Xanomeline | 131986-45-3 | MFCD00867179 | 100.0% | 281.42 g/mol | C14H23N3OS | 5 MG
SDP
Catalog No. 5000243784
Encompass_Preferred
Xanomeline is a selective muscarinic M1/M4 receptor agonist (CAS 131986-45-3) supplied as a solid analytical standard for laboratory research. It is provided in small pack sizes for analytical and preclinical studies; reported molecular formula C14H23N3OS and molecular weight 281.42 g/mol. Not for clinical use.
  • High purity suitable for analytical work.
  • Selective M1/M4 receptor agonist for neuroscience research.
  • Solid form enables easy weighing and storage.
  • Available in small package sizes for method development and pilot studies.
  • Accompanied by safety documentation for proper handling.
Catalog No. 50-002-43784 Supplier Medchemexpress LLC Supplier No. HY1051825MG
Xanomeline is a selective muscarinic M1/M4 receptor agonist (CAS 131986-45-3) supplied as a solid analytical standard for laboratory research. It is provided in small pack sizes for analytical and preclinical studies; reported molecular formula C14H23N3OS and molecular weight 281.42 g/mol. Not for clinical use.
  • High purity suitable for analytical work.
  • Selective M1/M4 receptor agonist for neuroscience research.
  • Solid form enables easy weighing and storage.
  • Available in small package sizes for method development and pilot studies.
  • Accompanied by safety documentation for proper handling.