Learn More
Medchemexpress LLC Xanomeline | 131986-45-3 | MFCD00867179 | 99.95% | 281.42 g·mol⁻¹ | C14H23N3OS | 10 MG
SDP
Catalog No. 5000243664
Encompass_Preferred
Xanomeline is a selective muscarinic M1/M4 receptor agonist used as a research reagent in neuroscience to investigate cholinergic signaling and neuropsychiatric disorders. It is provided as a high-purity chemical for laboratory use; consult the product data sheet and safety data sheet for handling, storage, and experimental guidance.
  • Selective muscarinic M1/M4 receptor agonist.
  • High purity suitable for analytical and biological research.
  • Available in multiple small-scale quantities for laboratory use.
  • Supplied with a product data sheet and a safety data sheet.
  • Used to study cholinergic signaling and neuropsychiatric disease models.
  • Compatible with in vitro and in vivo experimental protocols.
Catalog No. 50-002-43664 Supplier Medchemexpress LLC Supplier No. HY10518210MG
Xanomeline is a selective muscarinic M1/M4 receptor agonist used as a research reagent in neuroscience to investigate cholinergic signaling and neuropsychiatric disorders. It is provided as a high-purity chemical for laboratory use; consult the product data sheet and safety data sheet for handling, storage, and experimental guidance.
  • Selective muscarinic M1/M4 receptor agonist.
  • High purity suitable for analytical and biological research.
  • Available in multiple small-scale quantities for laboratory use.
  • Supplied with a product data sheet and a safety data sheet.
  • Used to study cholinergic signaling and neuropsychiatric disease models.
  • Compatible with in vitro and in vivo experimental protocols.