Learn More
Medchemexpress LLC Xanomeline 50mg | 131986-45-3 | 50MG
SDP
Catalog No. 5000244193
Encompass_Preferred
Xanomeline as an effective and selective muscarinic type 1 and type 4 (M1/M4) receptor agonist increases neuronal excitability Xanomeline can be used for the research of neurological disorders such as schizophrenia[1 [2
Catalog No. 50-002-44193 Supplier Medchemexpress LLC Supplier No. HY10518250MG
Xanomeline as an effective and selective muscarinic type 1 and type 4 (M1/M4) receptor agonist increases neuronal excitability Xanomeline can be used for the research of neurological disorders such as schizophrenia[1 [2