Learn More
Cayman Chemical Xanomeline (oxalate), Purity: ≥95%, CAS: 141064-23-5, 1 mg
SDP
Catalog No. 502887850
Encompass_Preferred
A potent agonist of muscarinic acetylcholine receptors (EC50 values are 0.3, 92.5, 5, 52, and 42 nM for M1, M2, M3, M4, and M5, respectively); has antipsychotic-like activities in rats and Cebus monkeys; enhances memory function and has utility in treating Alzheimer’s Disease
Catalog No. 50-288-7850 Supplier Cayman Chemical Supplier No. 107901 MG
A potent agonist of muscarinic acetylcholine receptors (EC50 values are 0.3, 92.5, 5, 52, and 42 nM for M1, M2, M3, M4, and M5, respectively); has antipsychotic-like activities in rats and Cebus monkeys; enhances memory function and has utility in treating Alzheimer’s Disease