Learn More
Medchemexpress LLC HY-13410 5mg Medchemexpress, Xanomeline (oxalate) CAS:141064-23-5 Purity:>98%
SDP
Catalog No. 502030306
Encompass_Preferred
Medchemexpress, HY-13410 5mg Xanomeline (oxalate) CAS:141064-23-5 Xanomeline oxalate (LY246708 oxalate) is a potent and selective muscarinic receptor agonist (SMRA) and stimulates phosphoinositide hydrolysis in vivo. Xanomeline oxalate can be used for the research of Alzheimer’s disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0306 Supplier Medchemexpress LLC Supplier No. HY134105MG
Medchemexpress, HY-13410 5mg Xanomeline (oxalate) CAS:141064-23-5 Xanomeline oxalate (LY246708 oxalate) is a potent and selective muscarinic receptor agonist (SMRA) and stimulates phosphoinositide hydrolysis in vivo. Xanomeline oxalate can be used for the research of Alzheimer’s disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.