Learn More
Abcam x anomeline o x alate, M1 muscarinic acetylcholine receptor agonist, 10MG
Catalog No. 502583372
Encompass_Preferred
MW 371.5 Da, Purity >99%. Functionally preferring M₁ muscarinic acetylcholine receptor agonist (pKi values are 7.1 (M₁), 6.9 (M₂), 7.4 (M₃), 7.7 (M₄) and 7.4 (M₅)). Able to cross blood-brain barrier. Displays antipsychotic activity.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3372 Supplier Abcam Supplier No. AB14107810MG
MW 371.5 Da, Purity >99%. Functionally preferring M₁ muscarinic acetylcholine receptor agonist (pKi values are 7.1 (M₁), 6.9 (M₂), 7.4 (M₃), 7.7 (M₄) and 7.4 (M₅)). Able to cross blood-brain barrier. Displays antipsychotic activity.
The product is subject to the following: Abcam Restricted Use Statement